AC5H5815FBDE Accela ChemBio Inc.

1-(Benzyloxy)-3-fluoro-2-methyl-4-nitrobenzene

  • MDL number MFCD31536676
  • Cas Number : 1020173-26-5
  • Molecular Weight : 261.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY059676&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY059676.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart