AC5H5815FBE4 Accela ChemBio Inc.

3,4-Dihydropyrazino[1,2-a]indol-1(2H)-one

  • MDL number MFCD08272112
  • Cas Number : 152193-85-6
  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY069069&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY069069.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart