AC5H5815FBED Accela ChemBio Inc.

Methyl 2-Amino-[1,2,4]triazolo[1,5-a]pyridine-6-carboxylate

  • MDL number MFCD11976480
  • Cas Number : 1094107-41-1
  • Molecular Weight : 192.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY102830&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY102830.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart