AC5H5815FBEE Accela ChemBio Inc.

4-(1-Hydroxyethyl)phenylboronic Acid

  • MDL number MFCD04112543
  • Cas Number : 518336-20-4
  • Molecular Weight : 165.98214

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY103371&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY103371.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart