AC5H5815FC01 Accela ChemBio Inc.

6-Bromo-8-nitro-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD31567136
  • Molecular Weight : 243.01762

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY129612&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY129612.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart