AC5H5815FC06 Accela ChemBio Inc.

2'-Methoxy-4'-nitrophenylAcetophenone

  • MDL number MFCD11847486
  • Cas Number : 90564-14-0
  • Molecular Weight : 195.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY216045&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY216045.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart