AC5H5815FC0D Accela ChemBio Inc.

2-(Isopropylamino)benzenethiol

  • MDL number MFCD20661572
  • Cas Number : 30459-53-1
  • Molecular Weight : 167.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY232861&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY232861.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart