AC5H5815FC0E Accela ChemBio Inc.

3,5-Dibromo-4-fluorobenzoic Acid

  • MDL number MFCD28784654
  • Cas Number : 402-87-9
  • Molecular Weight : 297.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY233054&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY233054.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart