AC5H5815FC0F Accela ChemBio Inc.

(S)-1-[2-(Boc-amino)acetyl]-4,4-difluoropyrrolidine-2-carboxamide

  • MDL number MFCD29049849
  • Cas Number : 1448440-48-9
  • Molecular Weight : 307.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY233674&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY233674.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart