AC5H5815FC13 Accela ChemBio Inc.

Methyl (R)-3-Boc-1,2,3-oxathiazolidine-4-carboxylate 2,2-Dioxide

  • MDL number MFCD31667701
  • Cas Number : 1621706-89-5
  • Molecular Weight : 281.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY235422&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY235422.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart