AC5H5815FC1D Accela ChemBio Inc.

4-(Benzyloxy)-8-methyl-2-naphthoic Acid

  • Molecular Weight : 292.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY249687&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY249687.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart