AC5H5815FC1E Accela ChemBio Inc.

N-Boc-4-(benzyloxy)-8-methyl-2-naphthalenamine

  • Molecular Weight : 363.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY249688&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY249688.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart