AC5H5815FC23 Accela ChemBio Inc.

7-Bromo-3,3-dimethyl-1,2,3,4-tetrahydronaphthalen-1-amine

  • MDL number MFCD20924977
  • Cas Number : 1782902-65-1
  • Molecular Weight : 254.16614

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY251003&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY251003.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart