AC5H5815FC24 Accela ChemBio Inc.

5-(3,5-Difluorophenyl)-4,5-dihydro-1H-pyrazole

  • MDL number MFCD31978008
  • Cas Number : 2049868-30-4
  • Molecular Weight : 182.1700264

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY251032&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY251032.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart