AC5H5815FC27 Accela ChemBio Inc.

2,6-Dibromo-4-fluorobenzyl Acetate

  • MDL number MFCD29054170
  • Cas Number : 1346674-62-1
  • Molecular Weight : 325.9570832

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY251501&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY251501.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart