AC5H5815FC28 Accela ChemBio Inc.

5-Bromo-4-fluoro-2-iodopyridine

  • MDL number MFCD18250717
  • Cas Number : 1260669-95-1
  • Molecular Weight : 301.8829532

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY252991&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY252991.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart