AC5H5815FC2B Accela ChemBio Inc.

2,3-Dihydro-1H-pyrrolo[3,2-b]pyridine Dihydrochloride

  • MDL number MFCD24843050
  • Cas Number : 1443981-64-3
  • Molecular Weight : 193.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to orange solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY253170&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY253170.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart