AC5H5815FC2D Accela ChemBio Inc.

1-Bromo-8-methylnaphthalene

  • MDL number MFCD00229485
  • Cas Number : 33295-37-3
  • Molecular Weight : 221.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY253837&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY253837.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart