AC5H5815FC2F Accela ChemBio Inc.

5-Fluoro-2-methoxy-4-(4-methyl-1-piperazinyl)aniline

  • MDL number MFCD32068339
  • Cas Number : 878155-86-3
  • Molecular Weight : 239.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY253961&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY253961.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart