AC5H5815FC30 Accela ChemBio Inc.

4-Amino-6-fluorocinnoline-3-carboxylic Acid

  • MDL number MFCD24395657
  • Cas Number : 161373-43-9
  • Molecular Weight : 207.1612432

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY254044&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY254044.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart