AC5H5815FC32 Accela ChemBio Inc.

Ethyl 3-Cyano-3-methylpentanoate

  • MDL number MFCD32068497
  • Cas Number : 39121-94-3
  • Molecular Weight : 169.2209

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY254099&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY254099.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart