AC5H5815FC34 Accela ChemBio Inc.

5-Chloro-3-methoxypyridazine

  • MDL number MFCD32177081
  • Molecular Weight : 144.559

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY257765&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY257765.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart