AC5H5815FC35 Accela ChemBio Inc.

Methyl 5-Chloro-4-iodo-1H-pyrrole-2-carboxylate

  • MDL number MFCD32177082
  • Cas Number : 903560-51-0
  • Molecular Weight : 285.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY257780&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY257780.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart