AC5H5815FC36 Accela ChemBio Inc.

Methyl 4-Bromo-5-chloro-1H-pyrrole-2-carboxylate

  • MDL number MFCD31638576
  • Cas Number : 1629633-64-2
  • Molecular Weight : 238.4664

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY257781&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY257781.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart