AC5H5815FC38 Accela ChemBio Inc.

Ethyl 1-(Cyanomethyl)-1H-indole-2-carboxylate

  • MDL number MFCD02928408
  • Cas Number : 126718-04-5
  • Molecular Weight : 228.24658

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY257803&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY257803.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart