AC5H5815FC3E Accela ChemBio Inc.

5-[(7-Chloro-4-quinolyl)amino]-2-[(diethylamino)methyl]phenol

  • Cas Number : 1643-45-4
  • Molecular Weight : 355.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY261673&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY261673.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart