AC5H5815FC40 Accela ChemBio Inc.

Phenyl 2-Methoxyacetate

  • MDL number MFCD08777398
  • Cas Number : 16839-92-2
  • Molecular Weight : 166.1739

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY261726&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY261726.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart