AC5H5815FC43 Accela ChemBio Inc.

Ethyl 6-Chlorobenzofuran-2-carboxylate

  • MDL number MFCD28987315
  • Cas Number : 74222-19-8
  • Molecular Weight : 224.64036

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY261964&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY261964.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart