AC5H5815FC46 Accela ChemBio Inc.

(R)-3-Phenylpiperidine Hydrochloride

  • MDL number MFCD08461140
  • Cas Number : 450416-58-7
  • Molecular Weight : 197.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3R)-3-phenylpiperidine hydrochloride
Inchi Code 1S/C11H15N.ClH/c1-2-5-10(6-3-1)11-7-4-8-12-9-11;/h1-3,5-6,11-12H,4,7-9H2;1H/t11-;/m0./s1
Inchi Key OKVMJYYCFLIGFJ-MERQFXBCSA-N

Description

Purity 95%
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY262091&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY262091.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart