AC5H5815FC4A Accela ChemBio Inc.

1-(1-Boc-4-piperidyl)-3-bromo-2-propanone

  • MDL number MFCD11975633
  • Cas Number : 473795-43-6
  • Molecular Weight : 320.22268

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY262174&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY262174.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart