AC5H5815FC4E Accela ChemBio Inc.

Trimethyl[4-(trifluoromethyl)phenyl]silane

  • MDL number MFCD22053136
  • Cas Number : 312-75-4
  • Molecular Weight : 218.2909296

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY262299&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY262299.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart