AC5H5815FC4F Accela ChemBio Inc.

N-[2-(Boc-amino)ethyl]-4-chloropyridine-2-carboxamide

  • MDL number MFCD32177094
  • Cas Number : 103878-75-7
  • Molecular Weight : 299.75332

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY262304&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY262304.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart