AC5H5815FC52 Accela ChemBio Inc.

(3-Nitrobenzyl)hydrazine Dihydrochloride

  • Molecular Weight : 240.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY262329&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY262329.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart