AC5H5815FC54 Accela ChemBio Inc.

1-(5-Fluoro-2-pyridyl)-1H-pyrazole-4-boronic Acid

  • MDL number MFCD31724620
  • Cas Number : 2256747-78-9
  • Molecular Weight : 206.9694832

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY262663&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY262663.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart