AC5H5815FC56 Accela ChemBio Inc.

3-(Methoxymethyl)-1H-pyrazole

  • MDL number MFCD20231592
  • Cas Number : 37599-33-0
  • Molecular Weight : 112.12982

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY262778&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY262778.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart