AC5H5815FC58 Accela ChemBio Inc.

3-Pentyn-2-one

  • MDL number MFCD19300617
  • Cas Number : 7299-55-0
  • Molecular Weight : 82.10054

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY262884&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY262884.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart