AC5H5815FC5A Accela ChemBio Inc.

4-Bromotetrahydrofuran-3-ol

  • MDL number MFCD20627320
  • Cas Number : 204926-49-8
  • Molecular Weight : 167.00118

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY262931&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY262931.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart