AC5H5815FC5B Accela ChemBio Inc.

Ethyl 4-Methylpiperazine-1-carboxylate

  • MDL number MFCD00030790
  • Cas Number : 59325-11-0
  • Molecular Weight : 172.22484

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY262998&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY262998.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart