AC5H5815FC5C Accela ChemBio Inc.

4-Chloro-3-cyano-2-fluorobenzoic Acid

  • MDL number MFCD28747740
  • Cas Number : 1628684-15-0
  • Molecular Weight : 199.5663232

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY263025&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY263025.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart