AC5H5815FC60 Accela ChemBio Inc.

6-Methyl-1-cyclohexene-1-boronic Acid Pinacol Ester

  • MDL number MFCD22494818
  • Cas Number : 448211-44-7
  • Molecular Weight : 222.13152

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY263139&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY263139.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart