AC5H5815FC64 Accela ChemBio Inc.

5-(2-Fluorophenyl)-1H-1,2,4-triazole

  • MDL number MFCD28397300
  • Cas Number : 1505273-47-1
  • Molecular Weight : 163.1517432

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY263341&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY263341.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart