AC5H5815FC65 Accela ChemBio Inc.

Ethyl 1-Fluoro-2-oxocyclohexanecarboxylate

  • MDL number MFCD01076477
  • Cas Number : 1578-70-7
  • Molecular Weight : 188.1961232

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY263469&attname=GoodsCode

Safety Information

pcodes P261-P280a-P305+P351+P338-P304+P340-P405-P501a
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY263469.pdf
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart