AC5H706BF05F Accela ChemBio Inc.

Ethyl alpha,alpha-Difluorophenylacetate

  • MDL number MFCD08275296
  • Cas Number : 2248-46-6
  • Molecular Weight : 200.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY038854&attname=GoodsCode

Safety Information

pcodes P261-P280a-P305+P351+P338-P304+P340-P405-P501a
MSDS http://pdf.accelachem.com/cn/msds/SY038854.pdf
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart