AC5H706BF0EF Accela ChemBio Inc.

N,N-Diethylcyclopropanecarboxamide

  • MDL number MFCD00760286
  • Cas Number : 10374-28-4
  • Molecular Weight : 141.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N,N-diethylcyclopropanecarboxamide
Inchi Code 1S/C8H15NO/c1-3-9(4-2)8(10)7-5-6-7/h7H,3-6H2,1-2H3
Inchi Key VZLNARVNYHVMHK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY069853&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY069853.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart