AC5H706BF341 Accela ChemBio Inc.

5-Methoxy-2,2-dimethyl-2H-chromene-6-carboxylic Acid

  • MDL number MFCD32632803
  • Cas Number : 62499-04-1
  • Molecular Weight : 234.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY264903&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY264903.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart