AC5H706BF366 Accela ChemBio Inc.

2-Bromo-1-cyclohexenecarbaldehyde

  • MDL number MFCD16657669
  • Cas Number : 38127-47-8
  • Molecular Weight : 189.04976

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY266273&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY266273.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart