AC5H93DE82BD Accela ChemBio Inc.

(2-Chloroethyl)sulfamoyl Chloride

  • MDL number MFCD12547701
  • Cas Number : 53627-11-5
  • Molecular Weight : 178.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloroethylsulfamoyl chloride
Inchi Code 1S/C2H5Cl2NO2S/c3-1-2-5-8(4,6)7/h5H,1-2H2
Inchi Key YKEZOKIPIIBNDN-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. -20?
Physical Form Colorless to yellow liquid
Shipping Temperature +4 ?
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018715&attname=GoodsCode

Safety Information

signalword Warning
MSDS http://pdf.accelachem.com/cn/msds/SY018715.pdf
hcodes GHS05, GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart