AC5H93DE82DD Accela ChemBio Inc.

4-Chloro-2-formylbenzonitrile

  • MDL number MFCD10699537
  • Cas Number : 77532-89-9
  • Molecular Weight : 165.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY037459&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY037459.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart