AC5H93DE82E0 Accela ChemBio Inc.

4-(2,2-Difluoroethoxy)phenylboronic Acid

  • MDL number MFCD11111926
  • Cas Number : 958453-65-1
  • Molecular Weight : 201.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2,2-difluoroethoxy)phenylboronic acid
Inchi Code 1S/C8H9BF2O3/c10-8(11)5-14-7-3-1-6(2-4-7)9(12)13/h1-4,8,12-13H,5H2
Inchi Key ZSNRGSJVRBZBET-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY038143&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY038143.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart