AC5H93DE82F0 Accela ChemBio Inc.

3'-Acetoxyacetophenone

  • MDL number MFCD00017227
  • Cas Number : 2454-35-5
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-acetylphenyl acetate
Inchi Code 1S/C10H10O3/c1-7(11)9-4-3-5-10(6-9)13-8(2)12/h3-6H,1-2H3
Inchi Key OTHYPAMNTUGKDK-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY051095&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY051095.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart